2-Chloro-3-picoline

Warning

Synonyms: 2-Chloro-3-methylpyridine, 2-Chloro 3-methylpyridine, 2-Chloro-β-picoline

518948-25ML 518948-5ML
220074-163EA 287.69 USD
220074-163 220074-164
2-Chloro-3-picoline
2-Chloro-3-picoline
Formula: C₆H₆ClN
MW: 127.57 g/mol
MDL Number: MFCD00234177
CAS Number: 18368-76-8
EINECS: 242-242-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR