2-Amino-5-fluoropyridine

518689-5G 518689-1G
220074-623EA 247.69 USD
220074-623 220074-622
2-Amino-5-fluoropyridine
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD01861120
CAS Number: 21717-96-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR