1-Heptanesulfonic acid sodium salt monohydrate ≥96.0% (by titrimetric analysis)

Synonyms: Sodium 1-heptanesulphonate monohydrate

51835-250G 51835-50G 51835-10G
220074-353EA 983.23 USD
220074-353 220074-354 220074-352
1-Heptanesulfonic acid sodium salt monohydrate ≥96.0% (by titrimetric analysis)
1-Heptanesulphonic acid sodium salt monohydrate
Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5

Order Now

Specification Test Results

Assay 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR