2-Chloro-5-(chloromethyl)pyridine

516910-5G 516910-25G
220074-558EA 50.83 USD
220074-558 220074-557
2-Chloro-5-(chloromethyl)pyridine
2-Chloro-5-(chloromethyl)pyridine
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR