4,4'-Diaminodiphenyl ether ≥97%

Synonyms: p-Aminophenyl ether, 4,4'-Oxydianiline, 4,4-Oxydianiline, Bis(4-aminophenyl) ether, 4-Aminophenyl ether

516805-100G 516805-500G
220074-548EA 68.49 USD
220074-548 220074-549
4,4'-Diaminodiphenyl ether ≥97%
4,4'-Diaminodiphenyl ether
Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Boiling Pt: 389 °C (1013 hPa)
Melting Pt: 184 °C
Flash Pt: 218 °C
MDL Number: MFCD00007863
CAS Number: 101-80-4
EINECS: 202-977-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR