Cyclopentyl isocyanate ≥97%

516589-5G 516589-1G
220074-536EA 75.38 USD
220074-536 220074-535
Cyclopentyl isocyanate ≥97%
Cyclopentyl isocyanate
Formula: C₆H₉NO
MW: 111.14 g/mol
MDL Number: MFCD02093695
CAS Number: 4747-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR