α-Bromo-4-iodotoulene ≥95%

Danger

Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

515604-1G 515604-5G
220074-436EA 37.69 USD
220074-436 220074-437
α-Bromo-4-iodotoulene ≥95%
α-Bromo-4-iodotoulene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 115 - 120 °C (0.6 -rr)
Melting Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR