2-Bromo-5-methylthiophene

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

515485-5G 515485-1G
220074-419EA 116.54 USD
220074-419 220074-418
2-Bromo-5-methylthiophene
2-Bromo-5-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.5529 g/cm³ (20 °C)
MDL Number: MFCD00130103
CAS Number: 765-58-2
EINECS: 212-151-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR