5-Bromo-o-toluidine ≥97%

Synonyms: 5-Bromo-2-methylaniline, 5-Bromo-o-toluidine, 2-Amino-4-bromotoluene

515019-25G 515019-5G
220073-872EA 54.23 USD
220073-872 220073-873
5-Bromo-o-toluidine ≥97%
5-Bromo-o-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 33 °C
Density: 1.49 g/cm³ (25 °C)
MDL Number: MFCD00800678
CAS Number: 39478-78-9
EINECS: 254-467-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR