Dibutyl sulfide

Synonyms: Dibutylsulphide, Butyl sulphide, n-Butyl sulphide

51491-500ML 51491-100ML
220074-341EA 359.69 USD
220074-341 220074-340
Dibutyl sulfide
Dibutyl sulphide
Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: -80 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00009468
CAS Number: 544-40-1
EINECS: 208-870-5
UN: 3334
ADR: 9,
Merck Index: 12,01626

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR