2,3-Dichlorophenylboronic acid

514047-25G 514047-5G
220074-262EA 165.38 USD
220074-262 220074-263
2,3-Dichlorophenylboronic acid
2,3-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR