2,3-Difluorophenylboronic acid

Synonyms: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

514039-25G 514039-5G
220074-260EA 215.38 USD
220074-260 220074-261
2,3-Difluorophenylboronic acid
2,3-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01863170
CAS Number: 121219-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR