2,5-Difluorophenylboronic acid

Synonyms: (2,5-Difluorophenyl)boronic acid, 2,5-Difluorobenzeneboronic acid, (2,5-Difluorophenyl)metaboric acid, Dihydroxy(2,5-difluorophenyl)borane

514020-25G 514020-5G
220074-258EA 86.15 USD
220074-258 220074-259
2,5-Difluorophenylboronic acid
2,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01863171
CAS Number: 193353-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR