2-Iodopyrazine

513164-1G 513164-5G
220074-117EA 51.58 USD
220074-117 220074-118
2-Iodopyrazine
2-Iodopyrazine
Formula: C₄H₃IN₂
MW: 205.99 g/mol
MDL Number: MFCD01319019
CAS Number: 32111-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR