3-Cyanophenylboronic acid

Warning

Synonyms: (3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid

513016-5G 513016-1G
220074-101EA 122.54 USD
220074-101 220074-100
3-Cyanophenylboronic acid
3-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
MW: 146.94 g/mol
MDL Number: MFCD01318967
CAS Number: 150255-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR