3-Pyridineboronic acid

Synonyms: 3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid

512125-5G 512125-25G 512125-1G
220073-990EA 347.54 USD
220073-990 220073-989 220073-988
3-Pyridineboronic acid
3-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR