2,6-Difluorophenyl isothiocyanate ≥98%

Synonyms: 2,6-Difluorophenyl isocyanate, 2,6-Difluorophenylisothiocyanate, 1,3-Difluoro-2-isothiocyanatobenzene

511781-5G 511781-1G
220073-948EA 80.77 USD
220073-948 220073-947
2,6-Difluorophenyl isothiocyanate ≥98%
2,6-Difluorophenyl isothiocyanate
Formula: C₇H₃F₂NS
MW: 171.17 g/mol
Boiling Pt: 106 - 107 °C (35 -rr)
Storage Temperature: Refrigerator
MDL Number: MFCD00041047
CAS Number: 207974-17-2
UN: 2930
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR