m-Toluidine ≥99%

Danger

Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline

511218-5ML 511218-100ML 511218-1L
220073-484EA 53.74 USD
220073-484 220073-864 220073-865
m-Toluidine ≥99%
m-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.98444 g/cm³ (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007808
CAS Number: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1,II
Merck Index: 13,09614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR