Methyl indole-5-carboxylate

Synonyms: Methyl 1H-indole-5-carboxylate

511188-5G 511188-1G
220073-859EA 84.32 USD
220073-859 220073-858
Methyl indole-5-carboxylate
Methyl indole-5-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00153023
CAS Number: 1011-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR