3,4,5-Trifluorophenol ≥98.0%

51092-5G-F 51092-1G-F
220072-837EA 546.32 USD
220072-837 220072-836
3,4,5-Trifluorophenol ≥98.0%
3,4,5-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042427
CAS Number: 99627-05-1
EINECS: 436-120-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR