Tantalum pentachloride, anhydrous

510688-5G 510688-25G
220073-838EA 155.38 USD
220073-838 220073-837
Tantalum pentachloride, anhydrous
Tantalum pentachloride
Formula: Cl₅Ta
Boiling Pt: 232…235 °C (1013 hPa)
Melting Pt: 216…217 °C
Density: 3.68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR