1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®

Synonyms: 1-Ethyl-3-methylimidazolium Acetate

51053-1KG-F 51053-100G-F
220073-822EA 1380 USD
220073-822 220073-821
1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium acetate
Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
MDL Number: MFCD06798186
CAS Number: 143314-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR