5-Bromo-2-methoxypyridine

Warning

510297-5ML 510297-25ML
220073-793EA 546.32 USD
220073-793 220073-792
5-Bromo-2-methoxypyridine
5-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR