tetra-Chloroauric(III) acid hydrate

Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

50790-1G 50790-5G 50790-250MG
220073-721EA 373.85 USD
220073-721 220073-723 220073-722
tetra-Chloroauric(III) acid hydrate
tetra-Chloroauric(III) acid hydrate
Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
Density: 1.169 g/cm³ (20 °C)
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay ~50% Au basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR