Methyl-3-bromobenzoate ≥98%

Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester

499625-25G 499625-5G
220072-277EA 67.31 USD
220072-277 220072-278
Methyl-3-bromobenzoate ≥98%
Methyl-3-bromobenzoate
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 69 - 70 °C (0.2 -rr)
Melting Pt: 31 °C
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR