1-Benzyl-3-methylimidazolium chloride

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium chloride

49914-5G-F 49914-50G-F
220072-744EA 61.61 USD
220072-744 220072-743
1-Benzyl-3-methylimidazolium chloride
1-Benzyl-3-methylimidazolium chloride
Formula: C₁₁H₁₃ClN₂
MW: 208.69 g/mol
MDL Number: MFCD07784451
CAS Number: 36443-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR