Calcium hydride ≥99.99%

Danger

Synonyms: Calcium dihydride

497355-10G 497355-2G
220071-648EA 1521.54 USD
220071-648 220071-649
Calcium hydride ≥99.99%
Calcium hydride
Formula: H₂Ca
MW: 42.09 g/mol
Melting Pt: 816 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I
Merck Index: 13,01674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR