Cobalt(II) thiocyanate

Warning

Synonyms: Cobalt dirhodanide, Cobaltous rhodanide, Cobalt(II) thiocyanate, Cobalt dithiocyanate, Cobaltous thiocyanate

497274-5G 497274-25G
220073-089EA 141.08 USD
220073-089 220073-088
Cobalt(II) thiocyanate
Cobalt (II) thiocyanate
Formula: C₂CoN₂S₂
MW: 175.1 g/mol
Density: 1.955 g/cm³ (25 °C)
MDL Number: MFCD00010937
CAS Number: 3017-60-5
EINECS: 221-156-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR