1-Methyl-3-propylimidazolium iodide

Synonyms: 1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide

49637-5G 49637-50G
220073-276EA 59.8 USD
220073-276 220073-275
1-Methyl-3-propylimidazolium iodide
1-Methyl-3-propylimidazolium iodide
Formula: C₇H₁₃IN₂
MW: 252.1 g/mol
MDL Number: MFCD08276385
CAS Number: 119171-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR