Tetrabromosilane ≥99.995%

Danger

Synonyms: Silicon tetrabromide, Silicon (IV) bromide

494100-25ML 494100-5ML
220073-044EA 486.15 USD
220073-044 220073-045
Tetrabromosilane ≥99.995%
Tetrabromosilane
Formula: Br₄Si
MW: 347.7 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: 5 °C
Density: 2.812 g/cm³ (25 °C)
MDL Number: MFCD00049530
CAS Number: 7789-66-4
EINECS: 232-182-4
UN: 3264
ADR: 8,II
Merck Index: 12,08643

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR