3-Methyloxindole

Synonyms: 3-Methyl-2-oxindole

493937-25G 493937-5G
220073-031EA 347.85 USD
220073-031 220073-032
3-Methyloxindole
3-Methyloxindole
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD04037370
CAS Number: 1504-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR