4-Cyanophenyl isocyanate ≥97%

493783-2G 493783-10G
220073-019EA 149.69 USD
220073-019 220073-018
4-Cyanophenyl isocyanate ≥97%
4-Cyanophenyl isocyanate
Formula: C₈H₄N₂O
MW: 144.13 g/mol
Boiling Pt: 123 °C (12 mmHg)
Melting Pt: 99 °C
CAS Number: 40465-45-0
EINECS: 254-934-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR