Indium nitride ≥99.9%

490628-1G 490628-5G
220072-855EA 375.38 USD
220072-855 220072-856
Indium nitride ≥99.9%
Indium nitride
Formula: InN
MW: 128.83 g/mol
Melting Pt: 1100 °C
Density: 6.73 to 6.88 g/cm³ (20 °C)
CAS Number: 25617-98-5
EINECS: 247-130-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR