(Trifluoromethyl)trimethylsilane

Synonyms: trimethyl(trifluoromethyl)silane

488712-25ML 488712-5ML
220072-602EA 378 USD
220072-602 220072-603
(Trifluoromethyl)trimethylsilane
(Trifluoromethyl)trimethylsilane
Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 55.5 °C (1013 hPa)
Density: 0.95 g/cm³ (20 °C)
MDL Number: MFCD00145454
CAS Number: 81290-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR