Octyl-3,4,5-trihydroxybenzoate

Warning

Synonyms: Octyl 3,4,5-trihydroxybenzoate, Gallic acid octyl ester, Octyl gallate, n-Octyl gallate

48700-250G-F 48700-50G-F
220072-441EA 625.69 USD
220072-441 220072-442
Octyl-3,4,5-trihydroxybenzoate
Octyl-3,4,5-trihydroxybenzoate
Formula: C₁₅H₂₂O₅
MW: 282.34 g/mol
Melting Pt: 95 - 100 °C
MDL Number: MFCD00002197
CAS Number: 1034-01-1
EINECS: 213-853-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR