Dodecyl-3,4,5-trihydroxybenzoate

Warning

Synonyms: Gallic acid lauryl ester, n-Dodecyl gallate, Dodecyl 3,4,5-trihydroxybenzoate, Gallic acid dodecyl ester, Dodecyl gallate, Lauryl 3,4,5-trihydroxybenzoate, Lauryl gallate, Progalline LA

48660-250G 48660-50G
220072-419EA 470.62 USD
220072-419 220072-420
Dodecyl-3,4,5-trihydroxybenzoate
Dodecyl-3,4,5-trihydroxybenzoate
Formula: C₁₉H₃₀O₅
MW: 338.44 g/mol
Melting Pt: 94…97 °C
MDL Number: MFCD00002195
CAS Number: 1166-52-5
EINECS: 214-620-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR