Ethyl-3,4,5-trihydroxybenzoate

Synonyms: Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate

48640-500G-F 48640-100G-F
220072-409EA 337.08 USD
220072-409 220072-408
Ethyl-3,4,5-trihydroxybenzoate
Ethyl-3,4,5-trihydroxybenzoate
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
MDL Number: MFCD00016430
CAS Number: 831-61-8
EINECS: 212-608-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR