2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.5%

Danger

Synonyms: Propylene glycol 1-methyl ether 2-acetat, 2-(1-Methoxy)propyl acetate, 1-Methoxy-2-propyl acetate, Propylene glycol methyl ether acetat, 1,2-Propanediol monomethyl ether acetate, DOWANOL PMA

484431-18L 484431-1L 484431-4L
220072-158EA 395.38 USD
220072-158 220072-159 220072-160
2-Methoxy-1-methylethyl acetate (PGMEA) ≥99.5%
2-Methoxy-1-methylethyl acetate (PGMEA)
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 145.8 (1013 hPa)
Melting Pt: -67 °C
Density: 0.968 g/cm³ (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00038500
CAS Number: 108-65-6
EINECS: 203-603-9
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR