Tetramethylammonium hydroxide pentahydrate

Danger

87741-25G 87741-500G 87741-100G
220098-985EA 49.71 USD
220098-985 220098-986 220098-984
Tetramethylammonium hydroxide pentahydrate
Tetramethylammonium hydroxide pentahydrate
Formula: C₄H₁₃NO·5H₂O
Melting Pt: 65 - 68 °C
Density: 1.829 g/cm³ (20 °C)
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR