(±)-1-Methoxy-2-propanol ≥99.5%

Danger

Synonyms: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol

484407-1L 484407-4L 484407-18L
220071-914EA 13.32 USD
220071-914 220072-155 220071-913
(±)-1-Methoxy-2-propanol ≥99.5%
(±)-1-Methoxy-2-propanol
Formula: CH₃CH(OH)CH₂OCH₃
MW: 90.12 g/mol
Boiling Pt: 119.6 °C (1013 hPa)
Melting Pt: -97 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR