Germanium(IV) oxide ≥99.99%

Warning

Synonyms: Germanium dioxide

483702-5G 483702-25G
220072-116EA 137.85 USD
220072-116 220072-115
Germanium(IV) oxide ≥99.99%
Germanium(IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1110 °C
Density: 3.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR