2,5-Dimethylphenylboronic acid

483516-25G 483516-5G
220072-099EA 22.92 USD
220072-099 220072-100
2,5-Dimethylphenylboronic acid
2,5-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01863525
CAS Number: 85199-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR