2,5-Dimethoxyphenylboronic acid

Synonyms: (2,5-Dimethoxyphenyl)metaboric acid, 2,5-Dimethoxybenzeneboronic acid, (2,5-Dimethoxyphenyl)boronic acid, Dihydroxy(2,5-dimethoxyphenyl)borane

483494-25G 483494-5G
220072-096EA 22.92 USD
220072-096 220072-097
2,5-Dimethoxyphenylboronic acid
2,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318181
CAS Number: 107099-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR