2,4-Dimethoxyphenylboronic acid ≥95%

483486-10G 483486-1G
220072-094EA 235.38 USD
220072-094 220072-095
2,4-Dimethoxyphenylboronic acid ≥95%
2,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01074590
CAS Number: 133730-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR