Gallium nitride ≥99.99%

Synonyms: Gallium nitride, Gallium(III) nitride

481769-50G 481769-10G
220071-968EA 1440 USD
220071-968 220071-967
Gallium nitride ≥99.99%
Gallium nitride
Formula: GaN
MW: 83.73 g/mol
Melting Pt: 800 °C
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00016108
CAS Number: 25617-97-4
EINECS: 247-129-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR