Hexaamminecobalt trichloride

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

481521-100G 481521-25G
220071-905EA 558.46 USD
220071-905 220071-952
Hexaamminecobalt trichloride
Hexaamminecobalt trichloride
Formula: H₁₈CoCl₃N₆
MW: 267.47 g/mol
Density: 1.702 g/cm³ (20 °C)
MDL Number: MFCD00036304
CAS Number: 10534-89-1
EINECS: 234-103-9
Merck Index: 12,04711

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR