Gold(I) chloride ≥99.9%

481130-5G 481130-1G
220070-340EA 1676.92 USD
220070-340 220070-101
Gold(I) chloride ≥99.9%
Gold(I) chloride
Formula: AuCl
MW: 232.42 g/mol
Melting Pt: 170 °C
Density: 7.57 g/cm³ (20 °C)
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR