2-Naphthyl boronic acid

Warning

Synonyms: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

480134-25G 480134-5G
220071-815EA 36.83 USD
220071-815 220071-816
2-Naphthyl boronic acid
2-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR