3,4-Dimethoxyphenylboronic acid

480118-25G 480118-5G
220071-334EA 59.54 USD
220071-334 220071-335
3,4-Dimethoxyphenylboronic acid
3,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01074574
CAS Number: 122775-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR