2,6-Dimethoxyphenylboronic acid

480096-25G 480096-5G
220071-332EA 39.46 USD
220071-332 220071-333
2,6-Dimethoxyphenylboronic acid
2,6-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR